missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lab Armor™ Walkabout™ Insulative Scoop Tray
Durable, light weight, small size for use under the hood or tight benchtop spaces
$205.57 - $515.41
Specifications
Capacity (Metric) | 0.5 L |
---|---|
Color | Black |
Height (Metric) | 15.24 cm |
Length (Metric) | 7.6 cm |
Material | PVC |
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
10876026
|
Lab Armor™
39438002 |
Tray only |
Each for $205.57
|
|
|||||
10-876-025
|
Lab Armor™
39438001 |
Tray Plus Beads |
Each for $515.41
|
|
|||||
Description
Durable, light weight, small size for use under the hood or tight benchtop spaces
It keeps the samples and reagents at temperature under hood, bench, after removing from bead bath, Chill bucket™, refrigerator or while traveling between labs.
- Molded one-piece expanded PVC construction
- Compatible for -196° to 93°C temperature
- Condensation free
- Can easily wash and disinfect
- For any size and shape vessels including non-water-tight vessels
- Dimension: 6 × 6 ×3 in. (15.24 x 15.24 x 7.6 cm)
- Capacity: 0.5L
Specifications
0.5 L | |
15.24 cm | |
PVC | |
15.24 cm | |
6 in. | |
6 in. |
Black | |
7.6 cm | |
Tray | |
3 in. | |
Rectangular |