missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lab Armor™ StayTemp™ Equipment Tray
For maintaining cold or heat with high efficiency in refrigerators, incubators, & ovens
Supplier: Lab Armor™ 39956001
Description
StayTemp Equipment trays are an upgrade that keeps samples and reagents in your refrigerator, incubator, and laboratory ovens at temperature despite normal hourly fluctuations in air temperature.
- Stainless steel construction
- Keeps samples organized protecting sample from defrost cycles and constant door opening and closing
- Eliminates the need of racks
- Can be hand washed, autoclaved or towel dried
- For any size and shape vessel including non-water-tight vessels
- Available in 3 sizes
Does not include lid.
Specifications
2 L | |
30.48 cm | |
Equipment Tray | |
For maintaining cold or heat with high efficiency in refrigerators, incubators and ovens | |
12 in. | |
Rectangular | |
10 lb. | |
4.5 kg | |
Autoclavable |
10.16 cm | |
Stainless Steel | |
15.2 cm | |
2 L Lab Armor Beads | |
4 in. | |
356°F | |
180°C | |
6 in. |