Please login to your online account to display your discounted pricing

Lab Armor™ Bead Bath


For warming, thawing, incubating, chilling maintaining constant temperature

Manufacturer: lab armor  74309720

 View more versions of this product

Catalog No. 10-876-008

  • / Each
 

Description & Specifications

Specifications

Warranty 1 Year
Voltage 120/230V
Hertz 50/60Hz
Amperage 5A
Weight (Metric) 44.4kg
Includes Beads, Stainless Steel body and lid
Electrical Requirements 120/230V 50/60Hz
Model M720
Product Type Bead Bath
Ambient Temperature (Metric) 5 to 80 °C
Volume (Metric) 20L
Plug Type Type B
Wattage 500w