Please login to your online account to display your discounted pricing
Lab-Aids™ Using Chemical Reactions to Reduce Waste
Wastes from industrial processes contain toxic or valuable substances.
$139.05 - $139.05
Specifications
Includes Teacher's manual with MSDS; 28 student worksheets and guides; 13 Chemplates™ 12 aluminum washers; 12 iron washers; 12 zinc washers; Six drop-controlled; bottles of waste copper chloride solution (15mL); Six plastic spoons; One dropper; One transparency
View More Specs
Includes: 
  • Teacher's manual with MSDS
  • 28 student worksheets and guides
  • 13 Chemplates™
  • 12 aluminum washers
  • 12 iron washers
  • 12 zinc washers
  • Six drop-controlled bottles of waste copper chloride solution (15mL)
  • Six plastic spoons
  • One dropper
  • One transparency
Products
Catalog Number Mfr. No. Product Type Price Quantity    
S96772
fisher scientific
441S
Making and Interpreting Topographic Maps Base Kit Each for $139.05
Description & Specifications
Specifications
Includes Teacher's manual with MSDS; 28 student worksheets and guides; 13 Chemplates™ 12 aluminum washers; 12 iron washers; 12 zinc washers; Six drop-controlled; bottles of waste copper chloride solution (15mL); Six plastic spoons; One dropper; One transparency
Industrial waste can sometimes be reclaimed and then recycled or disposed of safely.
  • Reclaim copper from a liquid “waste” containing dissolved copper chloride
  • Measure the effectiveness of three different metal-replacement reactions
  • Compare the effectiveness, cost and safety information for each replacement reaction
  • Decide which approach would be the best to use for removing copper from the “waste”
  • For six groups