missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lab-Aids™ Rock Cycle Activity
Reusable game reinforces the concepts of the rock cycle.
Supplier: Lab-Aids™ 404S
Includes: Eight rock cycle activity boards; eight number cubes; 32 activity pieces; 32 student worksheets and guides; 32 geologist's notes; teacher's manual; and eight sets each of 20 igneous rock cards, 20 metamorphic rock cards, 20 sedimentary rock cards and nine Earth process cards
Description
- Record changes to igneous, sedimentary, and metamorphic rocks
- Share data to construct and evaluate a scientific model
- Student sheet relates game results to the rock cycle
- Contains eight complete sets so an entire class can play simultaneously
Order Info
Class size: 32 students.
Specifications
1 Teacher's Guide; 32 Student Worksheets and Guides; 32 Geologist's Notes; 8 Rock Cycle activity boards; 32 Activity pieces; 8 Number Cubes; 9 Earth Process cards; 8 Igneous Rock Cards, Set of 20; 8 Metamorphic Rock Cards, Set of 20; 8 Sedimentary Rock Cards, Set of 20 | |
The Rock Cycle Game | |
One to two approx. 50-minute class periods | |
32 Students (8 Groups) |