Please login to your online account to display your discounted pricing

Lab-Aids™ Organic Chemistry Molecular Models: Isomers


Seven activities to construct a number of structural and stereoisomers.

Manufacturer: fisher science education  133

 View more versions of this product

Catalog No. S94055

  • / Each
 

Description & Specifications

Specifications

Grade High school
Includes Black Carbon Plastic tetrahedral centers (6), blue Oxygen double-bonded atoms (3), green Chlorine atoms (4), white Hydrogen atoms (12), covalent bonds (16), student worksheets (36) and instruction manual
Quantity Enough for 36 students
Type Organic chemistry molecular model, Isomers
For Use With (Application) Direct the students to draw and construct a number of structural and stereo isomers