Please login to your online account to display your discounted pricing

Lab-Aids™ Organic Chemistry Molecular Models: Isomers

Seven activities to construct a number of structural and stereoisomers.

$112.05 - $112.05

Specifications

Type Organic chemistry molecular model, Isomers
For Use With (Application) Direct the students to draw and construct a number of structural and stereo isomers
Grade High school
Includes Black Carbon Plastic tetrahedral centers (6), blue Oxygen double-bonded atoms (3), green Chlorine atoms (4), white Hydrogen atoms (12), covalent bonds (16), student worksheets (36) and instruction manual
View More Specs
Includes:  Each set includes: black carbon plastic tetrahedral centers (6), blue oxygen double-bonded atoms (3), green chlorine atoms (4), white hydrogen atoms (12), covalent bonds (16), student worksheets (36) and an instruction manual
Products
Catalog Number Mfr. No. Quantity Price Quantity    

S94055

 
fisher science education
133
Enough for 36 students Each for $112.05
Description & Specifications

Specifications

Type Organic chemistry molecular model, Isomers
For Use With (Application) Direct the students to draw and construct a number of structural and stereo isomers
Grade High school
Includes Black Carbon Plastic tetrahedral centers (6), blue Oxygen double-bonded atoms (3), green Chlorine atoms (4), white Hydrogen atoms (12), covalent bonds (16), student worksheets (36) and instruction manual

  • Differentiate two different organic compounds with the same formula by their structures
  • Construct models of structural isomerism in hydrocarbons, alcohol and aromatic derivatives
  • Observe stereoisomerism by constructing models of cis, trans and optical isomers