Lab-Aids™ Organic Chemistry Molecular Models: Hydrocarbons
Models include alkanes, alkenes, alkynes and benzene, including nonbonding valences.
$127.95 - $127.95
Includes: Each set includes: black carbon plastic tetrahedral centers (6), white hydrogen plastic atoms (12), covalent plastic bonds (19), student worksheets and guides (36) and instructions
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S19466 Fisher Science Education
LAI131
Each for $127.95
Specifications
Description
  • Construct a composite of all linkages, regardless of compatibility, in their determination of molecular properties
  • Understand how certain elements can combine to form organic compounds