Please login to your online account to display your discounted pricing

Lab-Aids™ Molecules of Metabolism Kit: Understanding Digestion and Respiration


3-D approach to the study of glycolysis and the Krebs Cycle.

Manufacturer: fisher science education  520EL

 View more versions of this product

Catalog No. S90185A

  • / Each
 

Description & Specifications

Specifications

Class Size 1 group
Includes 120 hydrogen atoms, 60 carbon atoms, 60 oxygen atoms, double bond, 220 bonds, set of labels and arrows, mitochondrion poster, 30 student worksheets and guides, instruction manual
Duration One ∽50 minute class period
For Use With (Equipment) Molecules of metabolism kit
For Use With (Application) Kit provides enough material for one additional group of 4 students
Product Type Molecules Of Metabolism Kit