Please login to your online account to display your discounted pricing

Lab-Aids™ Molecules of Metabolism Kit: Understanding Digestion and Respiration

3-D approach to the study of glycolysis and the Krebs Cycle.

$81.95 - $119.35

Specifications

Class Size 1 group
Product Type Molecules Of Metabolism Kit
Duration One ∽50 minute class period
View More Specs
Includes: 
  • 120 hydrogen atoms (white)
  • 60 carbon atoms (black)
  • 60 oxygen atoms, double bond (blue)
  • 220 bonds (white tubes)
  • Set of labels and arrows
  • Mitochondrion poster
  • 30 student worksheets and guides
  • Instruction manual
Products
Catalog Number Mfr. No. Includes For Use With (Application) Price Quantity    

S90185

 
fisher science education
520
Teacher's guide with msds, 30 student worksheets and guides, set of model pieces containing-123 white hydrogen atoms, 69 black carbon atoms, 72 blue double bond oxygen atoms, 263 bonds, set of labels and arrows and mitochondrion poster Student will be able to see and appreciate how large molecules are converted to smaller molecules that the body can use, kreb's cycle is illustrated in this kit Each for $119.35

S90185A

 
fisher science education
520EL
120 hydrogen atoms, 60 carbon atoms, 60 oxygen atoms, double bond, 220 bonds, set of labels and arrows, mitochondrion poster, 30 student worksheets and guides, instruction manual Kit provides enough material for one additional group of 4 students Each for $81.95
Description & Specifications

Specifications

Class Size 1 group
Product Type Molecules Of Metabolism Kit
Duration One ∽50 minute class period

Construct a diorama of anaerobic respiration (glycolysis) and aerobic respiration (the Krebs Cycle) in a step-by-step evolution.

  • See how large molecules convert to smaller molecules that the body can use
  • Addresses cellular respiration by illustrating adenosine triphosphate (ATP) formation
  • Observe molecules and understand the role of individual atoms and enzymes
  • Reusable kit is teacher developed with a station approach in mind

Expansion Kit

  • Allows more students to work on this activity at the same time
  • Contains eight additional molecular model packages