missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lab-Aids™ Molecular Model Kits

Hands-on learning activities help to visualize, construct and manipulate the invisible structure of molecules.

Manufacturer:  Lab-Aids™ LAI1251

 View more versions of this product

Catalog No. S19461



Description

Description

Introduction to Molecular Models Kit:

  • Colorful with easy-to-assemble pieces; includes two- and three-dimensional models
  • Specially designed two-dimensional color-coded atoms introduce molecular models and electron structure of atoms
  • Progress to three-dimensional color-coded atoms to visualize the structure of a formula and make behavior more understandable
  • Contains instructions and full background information
  • Class size: 50 students
Molecular Model Kit:
  • Furthers student understanding of molecular properties
  • Three-dimensional model for greater realism
  • Illustrates molecular shapes and special properties
  • Class size: 24 students working in pairs
Individual-Use, Student Organic Molecular Model Set:
  • Economical set designed for teacher-directed activities in organic chemistry
  • All components easily assemble from color-coded plastic pieces
  • Excellent for individual student study of homology, isomerism, functional groups and nomenclature Flexible design of covalent electron bonds makes construction of double and triple bonds easy

Specifications

Specifications

Student directions and guide, 50 covalent electron bonds, 10 Carbon tetrahedral (black), centers, 2 Nitrogen tetrahedral (red), centers, 4 Oxygen tetrahedral (blue), centers, 2 Bromine single electron (blue-green), atom units, 4 Chlorine single electron (green), atom units, 28 Hydrogen single electron (dark green), atom units, 2 Iodine single electron (dark green), atom units, 2 Oxygen double electron (blue), atom units
Molecular model kit
Documents