Lab-Aids™ Investigating Photovoltaic Cells Kit
Lets students investigate transformation of energy from sunlight into electrical energy using photovoltaic cells.
$391.43 - $391.43
Includes:
  • 28 Student worksheet and guides
  • One Teacher's guide
  • Six Electric motors
  • Six Photovoltaic cells
  • One Transparency: sample procedure and data table
  • Six Red wire leads with clips
  • Six Black wire leads with clips
  • Six Cardboard sunshields
Products 1
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S98795 View Documents Lab-Aids™
220S
Each for $391.43
Description
Description
  • Developed by SEPUP (The Science Education for Public Understanding Program)
  • Electrical circuits provide a means of transferring electrical energy
  • Investigate solar energy as an example of a renewable energy source
  • Explore how to power a motor using the photovoltaic cells and investigate some factors that affect the cell's ability to transform light energy into electricity
  • Discuss the advantages and disadvantages of using solar energy
Note: If any of your classes have more students than this kit accommodates, you can obtain extra components. Each Enlarge-A-Lab kit contains enough materials for two additional student teams.