Lab-Aids™ Identification of Substances Kit
A thorough introduction to acid and bases, observe effects of acids and bases on indicator dyes, measure concentration of acids by titration of an 'unknown' and perform a neutralization experiment.
Supplier:  Lab-Aids™ LAI83
Catalog No. S19395
Includes: All materials needed to identify four unknowns, worksheets/guides and instructions with SDS
Description
Description
Specific positive and negative ions are often used to identify inorganic substances. In this kit, students are asked to identify common metallic and non-metallic ions in zinc, cadmium, antimony, barium, sulfate, sulfide, sulfite, and carbonate. Through the identification process, students learn that each substance has distinctive properties. As a closing activity, students conduct chemical tests on four unknown substances. All test results are safe, clear and colorful.
Accommodates 3 classes, each with 12 groups of 2 students. Includes teacher guide, student sheets, assessment questions and SDS for all chemicals included.
Specifications
Specifications
Identification of Substances Kit
Colorful qualitative analysis brings excitement to the lab
All materials needed to identify four unknowns, worksheets/guides and instructions with sds
12 Students