Lab-Aids™ Classifying Sedimentary, Igneous and Metamorphic Rocks
Introduce and explain rock formation, describe and identify different rock types, use data to identify an unknown rock type and apply earth science to a real-world scenario.
Supplier: Lab-Aids™ 403S
Includes:
- Six Igneous Samples: Two each of gabbro (purple), granite (light blue) and obsidian (white)
- Six Sedimentary Samples: Two each of coal (yellow), limestone (blue) and sandstone (black)
- 12 Metamorphic Samples: Two each of gneiss (red) and phyllite (brown); eight garnet schist (green)
- Supplies: 16 magnifying glasses, 32 students worksheets and guides, 32 reading guides, teacher's manual
Description
Why study rocks? This kit helps you answer this question while providing rock samples for study. A fictional scenario describes the alleged discovery of diamonds in a national forest by a pair of hikers. Students learn that certain natural resources are often found in association with certain rocks. A reading on the formation of igneous, sedimentary, and metamorphic rocks supplements student observations as they examine, describe, and group rock samples. Students will use this knowledge to identify the hikers rock and decide whether the hikers were likely to have seen diamonds. Step-by-step student worksheets guide students to work in pairs. The rock samples provided in this kit are 1x2x1 in. or larger (except hiker's rocks) and clearly show characteristic features of sedimentary, igneous and metamorphic rocks. This kit is complete for 32 students and is reusable.Order Info
Class size: 32 students.
Specifications
1 Teacher's Guide; 32 Student Worksheets and Guides; 16 Magnifying lenses; 2 Gabbro (igneous rock) (purple); 2 Coal (sedimentary rock) (yellow); 8 Garnet schist (metamorphic rock) (green); 2 Gneiss (metamorphic rock) (red); 2 Limestone (sedimentary rock) (blue); 2 Obsidian (igneous rock) (white); 2 Phyllite (metamorphic rock) (brown); 2 Sandstone (sedimentary rock) (black); 2 Granite (igneous rock) (light blue) | |
Classifying Sedimentary, Igneous, and Metamorphic Rocks | |
One to two approx. 50-minute class periods | |
32 Students (8 Groups) |