missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-(+)-Valinol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1130006

 View more versions of this product

Catalog No. AAL1130006



Description

Description

L-(+)-Valinol reacts with aldehydes and nitriles to form imines and oxazolines, respectively, for asymmetric synthesis. It is also used for preparation of 4-isopropyloxazolidinones, employed in stereocontrolled aldol and alkylation reactions, synthesis of chiral 2-thiazolidinethiones for aldol reactions, asymmetric alkylation of condensation product with keto acids, asymmetric reactions of its amidines and imines and oxidation of N-protected derivative to the aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Valinol reacts with aldehydes and nitriles to form imines and oxazolines, respectively, for asymmetric synthesis. It is also used for preparation of 4-isopropyloxazolidinones, employed in stereocontrolled aldol and alkylation reactions, synthesis of chiral 2-thiazolidinethiones for aldol reactions, asymmetric alkylation of condensation product with keto acids, asymmetric reactions of its amidines and imines and oxidation of N-protected derivative to the aldehyde.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
Specifications

Specifications

L-(+)-Valinol
1.4548
80°C to 81°C (8mmHg)
5 g
+17° (c=10 in Ethanol)
C5H13NO
1719137
Soluble in water.
CC(C)[C@H](N)CO
103.17
103.17
2026-48-4
0.926
30°C to 35°C
78°C (172°F)
Air sensitive
MFCD00064296
l-valinol, s-+-2-amino-3-methyl-1-butanol, 2s-2-amino-3-methylbutan-1-ol, l-+-valinol, h-valinol, s-2-amino-3-methylbutan-1-ol, s-2-amino-3-methyl-1-butanol, 1-butanol, 2-amino-3-methyl-, 2s, s-2-amino-3-methyl-butan-1-ol
NWYYWIJOWOLJNR-RXMQYKEDSA-N
(2S)-2-amino-3-methylbutan-1-ol
640993
97%

Chemical Identifiers

2026-48-4
103.17
NWYYWIJOWOLJNR-RXMQYKEDSA-N
640993
CC(C)[C@H](N)CO
C5H13NO
MFCD00064296
l-valinol, s-+-2-amino-3-methyl-1-butanol, 2s-2-amino-3-methylbutan-1-ol, l-+-valinol, h-valinol, s-2-amino-3-methylbutan-1-ol, s-2-amino-3-methyl-1-butanol, 1-butanol, 2-amino-3-methyl-, 2s, s-2-amino-3-methyl-butan-1-ol
(2S)-2-amino-3-methylbutan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-975-5

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only