missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Valine tert-butyl ester hydrochloride, 97%

Manufacturer:  Alfa Aesar™ H5620706

 View more versions of this product

Catalog No. AAH5620706



Description

Description

Applications
It is involved in the synthesis of enantiomerically pure amino acid ester isocyanates.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

L-Valine tert-butyl ester hydrochloride
C9H20ClNO2
3693622
AUIVQIHTTVPKFS-FJXQXJEOSA-N
tert-butyl (2S)-2-amino-3-methylbutanoate;hydrochloride
12490702
97%
+4.4° (c=2 in Water)
13518-40-6
MFCD00034863
h-val-obut.hcl
CC(C)C(C(=O)OC(C)(C)C)N.Cl
209.714
209.72
5g

Chemical Identifiers

13518-40-6
209.714
AUIVQIHTTVPKFS-FJXQXJEOSA-N
12490702
CC(C)C(C(=O)OC(C)(C)C)N.Cl
C9H20ClNO2
MFCD00034863
h-val-obut.hcl
tert-butyl (2S)-2-amino-3-methylbutanoate;hydrochloride
Documents