missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Valine, Multi-Compendial, U.S.P., J.T. Baker™
Supplier: Avantor J.T.Baker 209506
Specifications
L-Valine | |
C5H11NO2 | |
Poly Bottle | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
(2S)-2-amino-3-methylbutanoic acid | |
6287 | |
117.15 | |
1 kg |
72-18-4 | |
MFCD00064220 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
CC(C)[C@H](N)C(O)=O | |
117.15 | |
CHEBI:16414 | |
Multi-Compendial/USP | |
1L = 1.23kg |
Chemical Identifiers
72-18-4 | |
117.15 | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
6287 | |
(2S)-2-amino-3-methylbutanoic acid |
C5H11NO2 | |
MFCD00064220 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
CHEBI:16414 | |
CC(C)[C@H](N)C(O)=O |