missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals Inc L-Valine, >99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 10476080
Specifications
L-Valine | |
White Powder | |
C5H11NO2 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
CC(C)C(C(=O)O)N | |
117.148 | |
CHEBI:16414 | |
Solid | |
100g |
72-18-4 | |
>99% | |
MFCD00064220 | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
(2S)-2-amino-3-methylbutanoic acid | |
6287 | |
117.15 | |
1.23g/mL(L) |
Safety and Handling
EINECSNumber : 200-773-6
Recommended Storage : Store at Room Temperature (15° to 30°C).