Please login to your online account to display your discounted pricing

L-Valine, >99%, MP Biomedicals™


Quantity 25g

Manufacturer: mp biomedicals  10476025

 View more versions of this product

Catalog No. ICN10476025

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 117.15
Quantity 25g
EINECS Number 200-773-6
Physical Form Solid
Density 1.23g/mL(L)
Appearance White Powder
Chemical Name or Material L-Valine
Assay Percent Range >99%
Molecular Formula C5H11NO2
MDL Number MFCD00064220
CAS 72-18-4
Synonym L-2-Amin-3-methylbutanoic acid
Recommended Storage Store at Room Temperature (15° to 30°C).

Specifications

Chemical Name or Material L-Valine
Assay Percent Range >99%
Molecular Formula C5H11NO2
Formula Weight 117.15
MDL Number MFCD00064220
Synonym L-2-Amin-3-methylbutanoic acid
Accessories

Store at Room Temperature (15° to 30°C).