missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Valine, >99%, MP Biomedicals™

$87.50 - $87.50

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Synonym 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, h-val-oh, l-alpha-amino-beta-methylbutyric acid, l-valine, s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, s-valine, valine, valinum latin
PubChem CID 6287
ChEBI CHEBI:16414
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10476080
View Documents
Mp Biomedicals
10476080
100g Each for $87.50
 
Specifications

Chemical Identifiers

72-18-4
117.148
KZSNJWFQEVHDMF-BYPYZUCNSA-N
6287
(2S)-2-amino-3-methylbutanoic acid
C5H11NO2
MFCD00064220
2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, h-val-oh, l-alpha-amino-beta-methylbutyric acid, l-valine, s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, s-valine, valine, valinum latin
CHEBI:16414
CC(C)C(C(=O)O)N

Specifications

72-18-4
>99%
MFCD00064220
KZSNJWFQEVHDMF-BYPYZUCNSA-N
(2S)-2-amino-3-methylbutanoic acid
6287
117.15
1.23g/mL(L)
L-Valine
White Powder
C5H11NO2
2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, h-val-oh, l-alpha-amino-beta-methylbutyric acid, l-valine, s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, s-valine, valine, valinum latin
CC(C)C(C(=O)O)N
117.148
CHEBI:16414
Solid
100g
Safety and Handling

Safety and Handling

EINECSNumber : 200-773-6

Recommended Storage : Store at Room Temperature (15° to 30°C).