missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Valinamide hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H6377522

 View more versions of this product

Catalog No. AAH6377522



Description

Description

Applications
It is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity

Solubility
Soluble in methanol (50 mg/ ml-clear, colorless solution).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

L-Valinamide hydrochloride
C5H13ClN2O
4198583
Soluble in methanol (50mg/ ml-clear,colorless solution).
CC(C)C(C(=O)N)N.Cl
152.622
152.62
95%
100g
3014-80-0
MFCD00039085
2s-2-amino-3-methylbutanamide hydrochloride, 2s-2-amino-3-methylbutanamide, chloride, butanamide, 2-amino-3-methyl-, monohydrochloride, 2s, d-valinamide-hcl, h-d-val-nh2 hcl, h-val-nh2 hcl, h-val-nh2.hcl, l-valinamide hydrochloride, l-valine amide, l-valine amide hydrochloride
XFCNYSGKNAWXFL-WCCKRBBISA-N
(2S)-2-amino-3-methylbutanamide;hydrochloride
13689583
Solid
266°C to 270°C

Chemical Identifiers

3014-80-0
152.622
XFCNYSGKNAWXFL-WCCKRBBISA-N
13689583
CC(C)C(C(=O)N)N.Cl
C5H13ClN2O
MFCD00039085
2s-2-amino-3-methylbutanamide hydrochloride, 2s-2-amino-3-methylbutanamide, chloride, butanamide, 2-amino-3-methyl-, monohydrochloride, 2s, d-valinamide-hcl, h-d-val-nh2 hcl, h-val-nh2 hcl, h-val-nh2.hcl, l-valinamide hydrochloride, l-valine amide, l-valine amide hydrochloride
(2S)-2-amino-3-methylbutanamide;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents