missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Valinamide hydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6377514
Description
It is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity
Solubility
Soluble in methanol (50 mg/ ml-clear, colorless solution).
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
266°C to 270°C | |
3014-80-0 | |
MFCD00039085 | |
l-valinamide hydrochloride, h-val-nh2 hcl, h-val-nh2.hcl, l-valine amide hydrochloride, 2s-2-amino-3-methylbutanamide hydrochloride, 2s-2-amino-3-methylbutanamide, chloride, butanamide, 2-amino-3-methyl-, monohydrochloride, 2s, l-valine amide, h-d-val-nh2 hcl, d-valinamide-hcl | |
XFCNYSGKNAWXFL-WCCKRBBISA-N | |
(2S)-2-amino-3-methylbutanamide;hydrochloride | |
13689583 | |
95% | |
Solid |
L-Valinamide hydrochloride | |
C5H13ClN2O | |
4198583 | |
Soluble in methanol (50mg/ ml-clear,colorless solution). | |
Cl.CC(C)[C@H](N)C(N)=O | |
152.62 | |
152.62 | |
25 g |
Chemical Identifiers
3014-80-0 | |
152.62 | |
XFCNYSGKNAWXFL-WCCKRBBISA-N | |
13689583 | |
Cl.CC(C)[C@H](N)C(N)=O |
C5H13ClN2O | |
MFCD00039085 | |
l-valinamide hydrochloride, h-val-nh2 hcl, h-val-nh2.hcl, l-valine amide hydrochloride, 2s-2-amino-3-methylbutanamide hydrochloride, 2s-2-amino-3-methylbutanamide, chloride, butanamide, 2-amino-3-methyl-, monohydrochloride, 2s, l-valine amide, h-d-val-nh2 hcl, d-valinamide-hcl | |
(2S)-2-amino-3-methylbutanamide;hydrochloride |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only