missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Tyrosylglycine, 95%

$394.92 - $394.92

Chemical Identifiers

CAS 673-08-5
Formula Weight 238.24
MDL Number MFCD00037779
Beilstein 2142807
Synonym H-Tyr-Gly-OH
Chemical Name or Material L-Tyrosylglycine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6670503 Alfa Aesar™
H6670503
1g Each for $394.92
AAH66705MD Alfa Aesar™
H66705MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

673-08-5
MFCD00037779
H-Tyr-Gly-OH
238.24
2142807
L-Tyrosylglycine

Specifications

238.24
673-08-5
1g
H-Tyr-Gly-OH
95%
Solid
MFCD00037779
2142807
L-Tyrosylglycine

Safety and Handling

storageNote1 : 2° to 8°C