missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Tyrosine tert-Butyl Ester, TCI America™

Manufacturer:  TCI America T244625G

 View more versions of this product

Catalog No. T244625G



Specifications

Specifications

L-Tyrosine tert-Butyl Ester
140°C
16874-12-7
MFCD00042644
DIGHFXIWRPMGSA-NSHDSACASA-N
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
6950582
Crystal-Powder at 20°C
White-Yellow
25g
C13H19NO3
l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068
CC(C)(C)OC(=O)C(CC1=CC=C(C=C1)O)N
237.299
237.30
≥97.0% (HPLC,T)

Chemical Identifiers

16874-12-7
237.299
DIGHFXIWRPMGSA-NSHDSACASA-N
6950582
CC(C)(C)OC(=O)C(CC1=CC=C(C=C1)O)N
C13H19NO3
MFCD00042644
l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents