missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Tyrosine tert-butyl ester, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435420010

 View more versions of this product

Catalog No. AC435420010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L-Tyrosine tert-butyl ester
Glass bottle
White
1g
98.5% min. (HPLC)
2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, h-tyr-obut, h-tyr-otbu, l-tyrosine t-butyl ester, l-tyrosine tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, tert-butyl l-tyrosinate, tyrosine, 1,1-dimethylethyl ester
CC(C)(C)OC(=O)C(CC1=CC=C(C=C1)O)N
237.3
237.3
99%
Authentic
25
142.5°C to 143.8°C
16874-12-7
C13H19NO3
DIGHFXIWRPMGSA-NSHDSACASA-N
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
6950582
Powder

Chemical Identifiers

16874-12-7
237.3
2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, h-tyr-obut, h-tyr-otbu, l-tyrosine t-butyl ester, l-tyrosine tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, tert-butyl l-tyrosinate, tyrosine, 1,1-dimethylethyl ester
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
C13H19NO3
DIGHFXIWRPMGSA-NSHDSACASA-N
6950582
CC(C)(C)OC(=O)C(CC1=CC=C(C=C1)O)N
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 240-902-3

SDS Documents