missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Tyrosine tert-butyl ester, 98%, Thermo Scientific™
$106.48 - $411.62
Chemical Identifiers
CAS | 16874-12-7 |
---|---|
Molecular Formula | C13H19NO3 |
Molecular Weight (g/mol) | 237.30 |
MDL Number | MFCD00042644 |
InChI Key | DIGHFXIWRPMGSA-LDGXTIHJNA-N |
Synonym | l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068 |
PubChem CID | 6950582 |
IUPAC Name | tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate |
SMILES | CC(C)(C)OC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5632203
|
Thermo Scientific Chemicals
H5632203 |
1 g |
Each for $106.48
|
|
|||||
AAH5632206
|
Thermo Scientific Chemicals
H5632206 |
5 g |
Each for $411.62
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
16874-12-7 | |
237.30 | |
DIGHFXIWRPMGSA-LDGXTIHJNA-N | |
6950582 | |
CC(C)(C)OC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
C13H19NO3 | |
MFCD00042644 | |
l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068 | |
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate |
Specifications
141°C to 144°C | |
C13H19NO3 | |
2696011 | |
DIGHFXIWRPMGSA-LDGXTIHJNA-N | |
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate | |
6950582 | |
98% | |
L-Tyrosine tert-butyl ester |
16874-12-7 | |
MFCD00042644 | |
l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068 | |
CC(C)(C)OC(=O)[C@@H](N)CC1=CC=C(O)C=C1 | |
237.30 | |
237.29 | |
1 g |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 240-902-3
TSCA : No
Recommended Storage : Ambient temperatures