Alfa Aesar™ L-Tyrosine tert-butyl ester, 98%
Manufacturer:  Alfa Aesar™ H5632203
 View more versions of this product
Catalog No. AAH5632203
Specifications
Specifications
L-Tyrosine tert-butyl ester
C13H19NO3
2696011
DIGHFXIWRPMGSA-NSHDSACASA-N
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
6950582
98%
1g
16874-12-7
MFCD00042644
l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068
CC(C)(C)OC(=O)C(CC1=CC=C(C=C1)O)N
237.299
237.29
141°C to 144°C
Chemical Identifiers
16874-12-7
237.299
DIGHFXIWRPMGSA-NSHDSACASA-N
6950582
CC(C)(C)OC(=O)C(CC1=CC=C(C=C1)O)N
C13H19NO3
MFCD00042644
l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Documents