missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Tyrosine Hydrazide, TCI America™

Manufacturer:  TCI America T18205G

 View more versions of this product

Catalog No. T18205G



Specifications

Specifications

L-Tyrosine Hydrazide
5g
C9H13N3O2
l-tyrosine hydrazide, s-2-amino-3-4-hydroxyphenyl propanehydrazide, tyrosine, hydrazide, l, l-tyrosinehydrazide, l-tyrosine, hydrazide, 2s-2-amino-3-4-hydroxyphenyl propanehydrazide, l-tyrosinohydrazide, tyrosine hydrazide, tyrosine hydrazone, l-tyrosine hydrazide
C1=CC(=CC=C1CC(C(=O)NN)N)O
195.222
195.22
≥98.0% (HPLC,T)
White-Yellow
7662-51-3
MFCD00007609
MWIXENPCUPDSOS-QMMMGPOBSA-N
(2S)-2-amino-3-(4-hydroxyphenyl)propanehydrazide
72779
Crystal-Powder at 20°C

Chemical Identifiers

7662-51-3
195.222
MWIXENPCUPDSOS-QMMMGPOBSA-N
72779
C1=CC(=CC=C1CC(C(=O)NN)N)O
C9H13N3O2
MFCD00007609
l-tyrosine hydrazide, s-2-amino-3-4-hydroxyphenyl propanehydrazide, tyrosine, hydrazide, l, l-tyrosinehydrazide, l-tyrosine, hydrazide, 2s-2-amino-3-4-hydroxyphenyl propanehydrazide, l-tyrosinohydrazide, tyrosine hydrazide, tyrosine hydrazone, l-tyrosine hydrazide
(2S)-2-amino-3-(4-hydroxyphenyl)propanehydrazide
Safety and Handling

Safety and Handling

TSCA : Yes

Documents