missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Tyrosine Hydrazide 98.0+%, TCI America™
$331.79 - $331.79
Chemical Identifiers
CAS | 7662-51-3 |
---|---|
Molecular Formula | C9H13N3O2 |
Molecular Weight (g/mol) | 195.22 |
MDL Number | MFCD00007609 |
InChI Key | MWIXENPCUPDSOS-QMMMGPOBSA-N |
Synonym | l-tyrosine hydrazide, s-2-amino-3-4-hydroxyphenyl propanehydrazide, tyrosine, hydrazide, l, l-tyrosinehydrazide, l-tyrosine, hydrazide, 2s-2-amino-3-4-hydroxyphenyl propanehydrazide, l-tyrosinohydrazide, tyrosine hydrazide, tyrosine hydrazone |
PubChem CID | 72779 |
IUPAC Name | (2S)-2-amino-3-(4-hydroxyphenyl)propanehydrazide |
SMILES | NNC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
T182025G
|
TCI America
T182025G |
25 g |
Each for $331.79
|
|
|||||
Chemical Identifiers
7662-51-3 | |
195.22 | |
MWIXENPCUPDSOS-QMMMGPOBSA-N | |
72779 | |
NNC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
C9H13N3O2 | |
MFCD00007609 | |
l-tyrosine hydrazide, s-2-amino-3-4-hydroxyphenyl propanehydrazide, tyrosine, hydrazide, l, l-tyrosinehydrazide, l-tyrosine, hydrazide, 2s-2-amino-3-4-hydroxyphenyl propanehydrazide, l-tyrosinohydrazide, tyrosine hydrazide, tyrosine hydrazone | |
(2S)-2-amino-3-(4-hydroxyphenyl)propanehydrazide |
Specifications
7662-51-3 | |
C9H13N3O2 | |
25 g | |
MWIXENPCUPDSOS-QMMMGPOBSA-N | |
(2S)-2-amino-3-(4-hydroxyphenyl)propanehydrazide | |
72779 | |
≥98.0% (HPLC,T) | |
L-Tyrosine Hydrazide |
White-Yellow | |
MFCD00007609 | |
l-tyrosine hydrazide, s-2-amino-3-4-hydroxyphenyl propanehydrazide, tyrosine, hydrazide, l, l-tyrosinehydrazide, l-tyrosine, hydrazide, 2s-2-amino-3-4-hydroxyphenyl propanehydrazide, l-tyrosinohydrazide, tyrosine hydrazide, tyrosine hydrazone | |
NNC(=O)[C@@H](N)CC1=CC=C(O)C=C1 | |
195.22 | |
195.22 | |
Crystalline Powder |
Safety and Handling
TSCA : Yes