Please login to your online account to display your discounted pricing

L-Tyrosine Hydrazide 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material L-Tyrosine hydrazide
Name Note 98%
CAS 7662-51-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H13N3O2
Linear Formula 4-(HO)C6H4CH2CH(NH2)CONHNH2
Formula Weight 195.22
MDL Number MFCD00007609
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160260250

  SDS
thermo fisher scientific
160260250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystals
Assay ≥97.5%
Formula Weight 195.22
Quantity 25g
Color White
EINECS Number 231-631-1
TSCA TSCA
Beilstein 14, I, 665
Fieser 02,454; 03,330
Chemical Name or Material L-Tyrosine hydrazide
Name Note 98%
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C9H13N3O2
Linear Formula 4-(HO)C6H4CH2CH(NH2)CONHNH2
Packaging Glass bottle
MDL Number MFCD00007609
CAS 7662-51-3