Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Tyrosine ethyl ester, 95%

Chemical Identifiers

Chemical Name or Material L-Tyrosine ethyl ester
CAS 949-67-7
Formula Weight 209.25
MDL Number MFCD00063046
Beilstein 2215181
Synonym H-Tyr-OEt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6635206

 
alfa aesar™
H6635206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6635214

 
alfa aesar™
H6635214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6635222

 
alfa aesar™
H6635222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 103° to 107°C
Formula Weight 209.25
Chemical Name or Material L-Tyrosine ethyl ester
Assay 95%
Synonym H-Tyr-OEt
TSCA TSCA
Physical Form Solid
Beilstein 2215181
CAS 949-67-7
Storage Note 1 -30° to -10°C
Quantity 5g
MDL Number MFCD00063046
Safety and Handling

-30° to -10°C