missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Tyrosine Benzyl Ester p-Toluenesulfonate, TCI America™

Manufacturer:  TCI America T24435G

 View more versions of this product

Catalog No. T24435G



Specifications

Specifications

L-Tyrosine Benzyl Ester p-Toluenesulfonate
177°C
53587-11-4
MFCD00038959
PJGVHBLZZQDFFM-RSAXXLAASA-N
benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid
104540
Crystal-Powder at 20°C
White
5g
C23H25NO6S
h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)C(CC2=CC=C(C=C2)O)N
443.514
443.51
≥98.0% (HPLC,T)

Chemical Identifiers

53587-11-4
443.514
PJGVHBLZZQDFFM-RSAXXLAASA-N
104540
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)C(CC2=CC=C(C=C2)O)N
C23H25NO6S
MFCD00038959
h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos
benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents