missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Tyrosine benzyl ester p-toluenesulfonate salt, 98%

Manufacturer:  Alfa Aesar™ H6336114

 View more versions of this product

Catalog No. AAH6336114



Description

Description

Applications
L-Tyrosine benzyl ester p-toluenesulfonate salt, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

L-Tyrosine benzyl ester p-toluenesulfonate salt
C23H25NO6S
3855339
PJGVHBLZZQDFFM-RSAXXLAASA-N
benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid
104540
Solid
25g
53587-11-4
MFCD00038959
h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)C(CC2=CC=C(C=C2)O)N
443.514
443.51
98%

Chemical Identifiers

53587-11-4
443.514
PJGVHBLZZQDFFM-RSAXXLAASA-N
104540
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)C(CC2=CC=C(C=C2)O)N
C23H25NO6S
MFCD00038959
h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos
benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents