missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Tyrosine benzyl ester p-toluenesulfonate salt, 98%, Thermo Scientific Chemicals
$439.74 - $439.74
Chemical Identifiers
CAS | 53587-11-4 |
---|---|
Molecular Formula | C23H25NO6S |
Molecular Weight (g/mol) | 443.51 |
MDL Number | MFCD00038959 |
InChI Key | PJGVHBLZZQDFFM-RSAXXLAASA-N |
Synonym | h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos |
PubChem CID | 104540 |
IUPAC Name | benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid |
SMILES | CC1=CC=C(C=C1)S(O)(=O)=O.N[C@@H](CC1=CC=C(O)C=C1)C(=O)OCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6336122
|
Thermo Scientific Chemicals
H6336122 |
100 g |
Each for $439.74
|
|
|||||
Description
L-Tyrosine benzyl ester p-toluenesulfonate salt, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Tyrosine benzyl ester p-toluenesulfonate salt, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
53587-11-4 | |
443.51 | |
PJGVHBLZZQDFFM-RSAXXLAASA-N | |
104540 | |
CC1=CC=C(C=C1)S(O)(=O)=O.N[C@@H](CC1=CC=C(O)C=C1)C(=O)OCC1=CC=CC=C1 |
C23H25NO6S | |
MFCD00038959 | |
h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos | |
benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid |
Specifications
53587-11-4 | |
MFCD00038959 | |
h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos | |
PJGVHBLZZQDFFM-RSAXXLAASA-N | |
benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid | |
104540 | |
98% | |
Solid |
C23H25NO6S | |
3855339 | |
Slightly soluble in water. | |
CC1=CC=C(C=C1)S(O)(=O)=O.N[C@@H](CC1=CC=C(O)C=C1)C(=O)OCC1=CC=CC=C1 | |
443.51 | |
443.51 | |
100 g | |
L-Tyrosine benzyl ester p-toluenesulfonate salt |
Safety and Handling
EINECSNumber : 258-650-8
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only