Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Tyrosine benzyl ester, 95%

Chemical Identifiers

Chemical Name or Material L-Tyrosine benzyl ester
CAS 52799-86-7
Formula Weight 271.32
MDL Number MFCD00191034
Beilstein 2901434
Synonym (S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propionate; H-Tyr-obzl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6440106

 
alfa aesar™
H6440106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6440114

 
alfa aesar™
H6440114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6440122

 
alfa aesar™
H6440122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 94° to 96°C
Formula Weight 271.32
Assay 95%
CAS 52799-86-7
Physical Form Solid
MDL Number MFCD00191034
Synonym (S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propionate; H-Tyr-obzl
Chemical Name or Material L-Tyrosine benzyl ester
Beilstein 2901434
Quantity 5g