missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Tryptophyl diphenylborinate, 95%

Manufacturer:  Alfa Aesar™ H5571203

 View more versions of this product

Catalog No. AAH5571203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L-Tryptophyl diphenylborinate
16655-49-5
MFCD22125148
XBYDPGDPEUHTPB-NRFANRHFSA-N
diphenylboranyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate
99738147
95%
1g
C23H21BN2O2
diphenylboranyl 2s-2-amino-3-1h-indol-3-yl propanoate
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC3=CNC4=CC=CC=C43)N
368.243
368.24

Chemical Identifiers

16655-49-5
368.243
XBYDPGDPEUHTPB-NRFANRHFSA-N
99738147
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CC3=CNC4=CC=CC=C43)N
C23H21BN2O2
MFCD22125148
diphenylboranyl 2s-2-amino-3-1h-indol-3-yl propanoate
diphenylboranyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents