missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Tryptophan (White Crystals or Cryst. Powder), Fisher BioReagents

Manufacturer:  Fisher BioReagents BP395100

Catalog No. BP395-100




Description

Description

This amino acid is suitable for use in tissue culture systems requiring additives.
Specifications

Specifications

L-Tryptophan
0.02% max.
10ppm max.
Pass Test
0.2% max.
Not Detected
Glass Bottle
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)N
-30 to -33° (+ or -)
204.229
CHEBI:16828
204.23
98.5 to 100.5%
White
5.5 to 7.0
73-22-3
1ppm max.
98.5 to 100.5 %
C11H12N2O2
15, 9977
ardeytropin, h-trp-oh, l-tryptophan, l-tryptophane, optimax, s-tryptophan, trofan, tryptacin, tryptophan, tryptophane
QIVBCDIJIAJPQS-VIFPVBQESA-N
0.1% max.
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
6305
95.0% min.
Powder/Solid
Tryptophan
280°C
100g

Chemical Identifiers

73-22-3
204.229
ardeytropin, h-trp-oh, l-tryptophan, l-tryptophane, optimax, s-tryptophan, trofan, tryptacin, tryptophan, tryptophane
CHEBI:16828
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)N
C11H12N2O2
QIVBCDIJIAJPQS-VIFPVBQESA-N
6305
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents
Product Certifications
Promotions

Promotions