missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Tryptophan Methyl Ester Hydrochloride, TCI America™

Manufacturer:  TCI America T165725G

 View more versions of this product

Catalog No. T165725G



Specifications

Specifications

L-Tryptophan Methyl Ester Hydrochloride
220°C
7524-52-9
MFCD00066134
XNFNGGQRDXFYMM-PPHPATTJSA-N
methyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
2734891
Crystal-Powder at 20°C
Undesignated
25g
C12H15ClN2O2
l-tryptophan methyl ester hydrochloride, h-trp-ome.hcl, methyl l-tryptophanate hydrochloride, methyl tryptophanate hydrochloride, methyl l-tryptophanate hcl, unii-au62o6861l, methyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, s-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, l-tryptophanmethylesterhydrochloride, l-tryptophan methyl ester hcl
COC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
254.714
254.71
≥98.0% (HPLC,T)

Chemical Identifiers

7524-52-9
254.714
XNFNGGQRDXFYMM-PPHPATTJSA-N
2734891
COC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
C12H15ClN2O2
MFCD00066134
l-tryptophan methyl ester hydrochloride, h-trp-ome.hcl, methyl l-tryptophanate hydrochloride, methyl tryptophanate hydrochloride, methyl l-tryptophanate hcl, unii-au62o6861l, methyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, s-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, l-tryptophanmethylesterhydrochloride, l-tryptophan methyl ester hcl
methyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents