Please login to your online account to display your discounted pricing

L-Tryptophan methyl ester hydrochloride, 98%, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC376650250

 View more versions of this product

Catalog No. AC376650250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 254.72
Physical Form Powder
Quantity 25g
Color White to off-white
Specific Rotation Condition + 18.00 (20.00°C c=5 CH3OH)
Assay Percent Range 98%
Molecular Formula C12H14N2O2·HCl
Specific Rotation +17° to +19° (20°C, 589nm) (c=5, CH3OH)
Packaging Plastic bottle
Additional Information Discolors on exposure to light
CAS 7524-52-9
Infrared Spectrum Authentic
Chemical Name or Material L-Tryptophan methyl ester hydrochloride
MDL Number MFCD00066134

Specifications

Chemical Name or Material L-Tryptophan methyl ester hydrochloride
CAS 7524-52-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H14N2O2·HCl
Formula Weight 254.72
MDL Number MFCD00066134