Please login to your online account to display your discounted pricing

L-Tryptophan methyl ester hydrochloride, 98%, ACROS Organics™


Quantity 5g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  AC376650050

 View more versions of this product

Catalog No. AC376650050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 254.72
Physical Form Powder
Quantity 5g
Color White to off-white
Specific Rotation Condition + 18.00 (20.00°C c=5 CH3OH)
Specific Rotation +17° to +19° (20°C, 589nm) (c=5, CH3OH)
MDL Number MFCD00066134
CAS 7524-52-9
Chemical Name or Material L-Tryptophan methyl ester hydrochloride
Molecular Formula C12H14N2O2·HCl
Packaging Plastic bottle
Additional Information Discolors on exposure to light
Infrared Spectrum Authentic
Assay Percent Range 98%

Specifications

Chemical Name or Material L-Tryptophan methyl ester hydrochloride
CAS 7524-52-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H14N2O2·HCl
Formula Weight 254.72
MDL Number MFCD00066134