missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Tryptophan methyl ester hydrochloride, 98%

$47.18 - $586.00

Chemical Identifiers

CAS 7524-52-9
Molecular Formula C12H15ClN2O2
Molecular Weight (g/mol) 254.714
MDL Number MFCD00066134
InChI Key XNFNGGQRDXFYMM-PPHPATTJSA-N
Synonym l-tryptophan methyl ester hydrochloride, h-trp-ome.hcl, methyl l-tryptophanate hydrochloride, methyl tryptophanate hydrochloride, methyl l-tryptophanate hcl, unii-au62o6861l, methyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, s-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, l-tryptophanmethylesterhydrochloride, l-tryptophan methyl ester hcl
PubChem CID 2734891
IUPAC Name methyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
SMILES COC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6260206 View Documents Alfa Aesar™
H6260206
5g Each for $47.18
 
AAH6260214 View Documents Alfa Aesar™
H6260214
25g Each for $168.09
 
AAH6260222 View Documents Alfa Aesar™
H6260222
100g Each for $586.00
 
Specifications

Chemical Identifiers

7524-52-9
254.714
XNFNGGQRDXFYMM-PPHPATTJSA-N
2734891
COC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
C12H15ClN2O2
MFCD00066134
l-tryptophan methyl ester hydrochloride, h-trp-ome.hcl, methyl l-tryptophanate hydrochloride, methyl tryptophanate hydrochloride, methyl l-tryptophanate hcl, unii-au62o6861l, methyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, s-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, l-tryptophanmethylesterhydrochloride, l-tryptophan methyl ester hcl
methyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride

Specifications

7524-52-9
MFCD00066134
l-tryptophan methyl ester hydrochloride, h-trp-ome.hcl, methyl l-tryptophanate hydrochloride, methyl tryptophanate hydrochloride, methyl l-tryptophanate hcl, unii-au62o6861l, methyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, s-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, l-tryptophanmethylesterhydrochloride, l-tryptophan methyl ester hcl
COC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
254.714
254.71
218°C to 220°C
L-Tryptophan methyl ester hydrochloride
C12H15ClN2O2
4240280
XNFNGGQRDXFYMM-PPHPATTJSA-N
methyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
2734891
98%
5g