missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Tryptophan Ethyl Ester Hydrochloride, TCI America™

Manufacturer:  TCI America T29815G

 View more versions of this product

Catalog No. T29815G



Specifications

Specifications

L-Tryptophan Ethyl Ester Hydrochloride
222°C
2899-28-7
MFCD00038993
PESYCVVSLYSXAK-MERQFXBCSA-N
ethyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
134519
Crystal-Powder at 20°C
Undesignated
5g
C13H17ClN2O2
l-tryptophan ethyl ester hydrochloride, ethyl l-tryptophanate hydrochloride, h-trp-oet.hcl, s-tryptophan ethyl ester hydrochloride, ethyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-trp-oet hcl, ethyl l-tryptophanate hcl, ethyl l-tryptophanate monohydrochloride, h-trp-oet. hcl, pubchem10920
CCOC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
268.741
268.74
≥95.0% (HPLC,T)

Chemical Identifiers

2899-28-7
268.741
PESYCVVSLYSXAK-MERQFXBCSA-N
134519
CCOC(=O)C(CC1=CNC2=CC=CC=C21)N.Cl
C13H17ClN2O2
MFCD00038993
l-tryptophan ethyl ester hydrochloride, ethyl l-tryptophanate hydrochloride, h-trp-oet.hcl, s-tryptophan ethyl ester hydrochloride, ethyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-trp-oet hcl, ethyl l-tryptophanate hcl, ethyl l-tryptophanate monohydrochloride, h-trp-oet. hcl, pubchem10920
ethyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents