Please login to your online account to display your discounted pricing

L-tryptophan ethyl ester hydrochloride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material L-Tryptophan ethyl ester hydrochloride
CAS 2899-28-7
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H16N2O2·HCl
Formula Weight 268.74
MDL Number MFCD00038993
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376670050

  SDS
fisher scientific acros
AC376670050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 268.74
Quantity 5g
Color Light gray
Assay Percent Range 99%
MDL Number MFCD00038993
Specific Rotation +9° to +11° (20°C, 589nm) (C=2, water)
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 2899-28-7
Chemical Name or Material L-Tryptophan ethyl ester hydrochloride
Molecular Formula C13H16N2O2·HCl