Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Tryptophan ethyl ester hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material L-Tryptophan ethyl ester hydrochloride
CAS 2899-28-7
Formula Weight 268.74
MDL Number MFCD00038993
Beilstein 3919010
Synonym H-Trp-OEt.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6695314

 
alfa aesar™
H6695314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6695322

 
alfa aesar™
H6695322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6695336

 
alfa aesar™
H6695336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 220° to 225°C
Formula Weight 268.74
Chemical Name or Material L-Tryptophan ethyl ester hydrochloride
Assay 97%
MDL Number MFCD00038993
Physical Form Powder
Beilstein 3919010
Quantity 25g
Color Pale brown
Synonym H-Trp-OEt.HCl
CAS 2899-28-7