Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Tryptophan benzyl ester hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material L-Tryptophan benzyl ester hydrochloride
CAS 35858-81-2
Formula Weight 330.81
MDL Number MFCD00058122
Synonym H-Trp-Obzl.HCl; (S)-Benzyl 2-amino-3-(3-indolyl)propionate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6338503

  SDS
alfa aesar™
H6338503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6338506

  SDS
alfa aesar™
H6338506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 330.81
Chemical Name or Material L-Tryptophan benzyl ester hydrochloride
Physical Form Powder
Assay 98%
MDL Number MFCD00058122
CAS 35858-81-2
Synonym H-Trp-Obzl.HCl; (S)-Benzyl 2-amino-3-(3-indolyl)propionate hydrochloride
Color White
Quantity 1g
Storage Note 1 2° to 8°C

Applications
L-Tryptophan benzyl ester hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol (50 mg/mL in MeOH).

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

2° to 8°C