missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Tryptophan, ≥99%, MP Biomedicals™
L-Tryptophan is an amino acid precursor of serotonin and melatonin
Supplier: Mp Biomedicals 0210315105
Specifications
L-Tryptophan | |
≥99% | |
MFCD00064340 | |
QIVBCDIJIAJPQS-VIFPVBQESA-N | |
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid | |
6305 | |
204.22 | |
White |
73-22-3 | |
C11H12N2O2 | |
ardeytropin, h-trp-oh, l-tryptophan, l-tryptophane, optimax, s-tryptophan, trofan, tryptacin, tryptophan, tryptophane | |
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)N | |
204.229 | |
CHEBI:16828 | |
Crystalline Powder | |
5g |
Chemical Identifiers
73-22-3 | |
204.229 | |
QIVBCDIJIAJPQS-VIFPVBQESA-N | |
6305 | |
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid |
C11H12N2O2 | |
MFCD00064340 | |
ardeytropin, h-trp-oh, l-tryptophan, l-tryptophane, optimax, s-tryptophan, trofan, tryptacin, tryptophan, tryptophane | |
CHEBI:16828 | |
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)N |
Safety and Handling
EINECSNumber : 200-795-6
Recommended Storage : Store at Room temperature (15 to 30°C).