Please login to your online account to display your discounted pricing

L-trans-2,4-PDC, Tocris

Transportable EAAT1-4 inhibitor/non-transportable EAAT5 inhibitor

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material L-trans-2, 4-PDC
Assay Percent Range >97%
Molecular Formula C6H9NO4
Formula Weight 159.14
Synonym L-trans-Pyrrolidine-2, 4-dicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

029810

 
r & d systems
029810
10mg Each for $195.49

029850

 
r & d systems
029850
50mg Each for $816.90
Description & Specifications

Specifications

Chemical Name or Material L-trans-2, 4-PDC
Quantity 10mg
CAS 64769-66-0
Recommended Storage Store at Room Temperature
Assay Percent Range >97%
Molecular Formula C6H9NO4
Formula Weight 159.14
Synonym L-trans-Pyrrolidine-2, 4-dicarboxylic acid
Safety and Handling

Store at Room Temperature