missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Threonyl diphenylborinate, 95%

Manufacturer:  Alfa Aesar™ H55599MD

 View more versions of this product

Catalog No. AAH55599MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L-Threonyl diphenylborinate
16655-44-0
MFCD22125157
YWSHJDZMGRKKKP-DOMZBBRYSA-N
diphenylboranyl (2S,3R)-2-amino-3-hydroxybutanoate
99738140
95%
250mg
C16H18BNO3
diphenylboranyl 2s,3r-2-amino-3-hydroxybutanoate
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(C(C)O)N
283.134
283.13

Chemical Identifiers

16655-44-0
283.134
YWSHJDZMGRKKKP-DOMZBBRYSA-N
99738140
B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(C(C)O)N
C16H18BNO3
MFCD22125157
diphenylboranyl 2s,3r-2-amino-3-hydroxybutanoate
diphenylboranyl (2S,3R)-2-amino-3-hydroxybutanoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents