L-threonine methyleester hydrochloride, +98%, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals 376690010
 View more versions of this product
Catalog No. AC376690010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
L-Threonine methyleester hydrochloride
98+%
Authentic
MFCD00037677
l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl
COC(=O)[C@@H]([NH3+])[C@@H](C)O
-9.5° to -10.5° (20°C, 589nm) (c=1, Methanol, 20°C)
2734893
≥98.0% ((on dry substance))
−10° (20°C c=1,methanol)
39994-75-7
C5H12NO3
CH3CH(OH)CH(NH2)C(O)OCH3·HCl
Glass bottle
TVHCXXXXQNWQLP-DMTCNVIQSA-O
methyl (2S,3R)-2-amino-3-hydroxybutanoate;hydrochloride
134.15
169.61
1% Max.
1g
Chemical Identifiers
39994-75-7
134.15
TVHCXXXXQNWQLP-DMTCNVIQSA-O
2734893
COC(=O)[C@@H]([NH3+])[C@@H](C)O
C5H12NO3
MFCD00037677
l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl
methyl (2S,3R)-2-amino-3-hydroxybutanoate;hydrochloride
SDS Documents