Please login to your online account to display your discounted pricing

L(-)-Thiazolidine-4-carboxylic Acid 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC138640100

 View more versions of this product

Catalog No. AC138640100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Flakes
Assay ≥97.5% (Acid)
Formula Weight 133.16
Quantity 10g
Color White to light yellow-beige to pink
RTECS Number XJ5425500
EINECS Number 252-106-3
Specific Rotation Condition −101° (20°C c=1,1N HCl)
Merck Index 09, 9040
Chemical Name or Material L(-)-Thiazolidine-4-carboxylic acid
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Packaging Glass bottle
Name Note 98%
Molecular Formula C4H7NO2S
MDL Number MFCD00005212
CAS 34592-47-7
Specific Rotation −136° to −144° (20°C, 589nm) (c=1, H2O)
Sulfated Ash 0.1% max.
Synonym Thioproline
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%

Specifications

Chemical Name or Material L(-)-Thiazolidine-4-carboxylic acid
Name Note 98%
CAS 34592-47-7
Assay ≥97.5% (Acid)
Assay Percent Range 98%
Molecular Formula C4H7NO2S
Formula Weight 133.16
MDL Number MFCD00005212
Synonym Thioproline
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents